Please login to your online account to display your discounted pricing

Rat anti-IL-2 R beta, Clone: TM-b1, Novus Biologicals™

$219.00 - $340.00

Specifications

Antigen IL-2 R beta
Clone TM-b1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Purified CD122.
Gene ID (Entrez) 3560
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14335501

 
novus biologicals canada
NBP14335501MG
0.1mg Each for $285.00

NBP14335505

 
novus biologicals canada
NBP143355005MG
0.05mg Each for $219.00

NBP1433555

 
novus biologicals canada
NBP14335505MG
0.5mg Each for $340.00
Description & Specifications

Specifications

Antigen IL-2 R beta
Clone TM-b1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Purified CD122.
Gene ID (Entrez) 3560
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Blocking
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Neutralization