Please login to your online account to display your discounted pricing

Rat anti-IL-2 R beta, Clone: TM-b1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143355005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14335505

  • / Each
 

Description & Specifications

Specifications

Applications Blocking
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Neutralization
Antigen IL-2 R beta
Formulation Phosphate buffer pH 7.2
Host Species Rat
Immunogen The immunogen for this antibody was Purified CD122.
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Mouse
Format Affinity Purified
Isotype IgG2b
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 3560
Clone TM-b1
Concentration 0.5mg/mL
Quantity 0.05mg