missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-IL-13, Clone: 13A, PE, Novus Biologicals™

$329.00 - $329.00

Specifications

Antigen IL-13
Clone 13A
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was IL13.
Gene ID (Entrez) 3596
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14372301 Novus Biologicals Canada
NBP14372301MG
0.1mg Each for $329.00
Specifications Description & Specifications

Specifications

IL-13
0.2mg/mL
3596
Monoclonal
Store at 4°C. Do not freeze.
IgG1, κ
Affinity Purified
Flow Cytometry
13A
The immunogen for this antibody was IL13.
PBS [pH 7.2]
Primary
Rat
PE
Affinity Purified