Please login to your online account to display your discounted pricing

Rat anti-IL-13, Clone: 13A, PE, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP143723005MG

 View all options for this product


For Research Use Only

Catalog No. NBP14372305


Description & Specifications


Antigen IL-13
Applications Flow Cytometry
Clone 13A
Concentration 0.2mg/mL
Conjugate PE
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Rat
Immunogen The immunogen for this antibody was IL13.
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3596