Please login to your online account to display your discounted pricing

Rat anti-IgD, Clone: 11-26c, PE, Novus Biologicals™

$131.00 - $329.00

Specifications

Antigen IgD
Clone 11-26c
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was IgD.
Formulation Aqueous buffer (contains stabilizer if necessary)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14373201

 
novus biologicals canada
NBP14373201MG
0.1mg Each for $252.00

NBP14373202

 
novus biologicals canada
NBP14373202MG
0.2mg Each for $329.00

NBP14373205

 
novus biologicals canada
NBP143732005MG
0.05mg Each for $131.00
Description & Specifications

Specifications

Antigen IgD
Clone 11-26c
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was IgD.
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2a, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry