Please login to your online account to display your discounted pricing

Rat anti-IgD, Clone: 11-26c, Novus Biologicals™

$219.00 - $307.00

Specifications

Antigen IgD
Clone 11-26c
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was IgD.
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14325801

 
novus biologicals canada
NBP14325801MG
0.1mg Each for $274.00

NBP14325805

 
novus biologicals canada
NBP143258005MG
0.05mg Each for $219.00

NBP1432585

 
novus biologicals canada
NBP14325805MG
0.5mg Each for $307.00
Description & Specifications

Specifications

Antigen IgD
Clone 11-26c
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was IgD.
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry