Please login to your online account to display your discounted pricing

Rat anti-IgD, Clone: 11-26c, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143258005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14325805

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Cross Reactivity Mouse
Monoclonal or Polyclonal Monoclonal
Purification Method Affinity Purified
Format Affinity Purified
Quantity 0.05mg
Isotype IgG2a, κ
Conjugate Unlabeled
Clone 11-26c
Immunogen The immunogen for this antibody was IgD.
Formulation Phosphate buffer pH 7.2
Host Species Rat
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Concentration 0.5mg/mL
Antigen IgD
Regulatory Status RUO