Please login to your online account to display your discounted pricing

Rat anti-IgD, Clone: 11-26c, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14325805MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1432585

  • / Each
 

Description & Specifications

Specifications

Antigen IgD
Applications Flow Cytometry
Clone 11-26c
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Rat
Immunogen The immunogen for this antibody was IgD.
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 0.5mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Murine