Please login to your online account to display your discounted pricing

Rat anti-IgD, Clone: 11-26c, Biotin, Novus Biologicals™

$131.00 - $296.00

Specifications

Antigen IgD
Clone 11-26c
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was IgD.
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP1435015

 
novus biologicals canada
NBP14350105MG
0.5mg Each for $296.00

NBP1435015M

 
novus biologicals canada
NBP143501005MG
0.05mg Each for $131.00
Description & Specifications

Specifications

Antigen IgD
Clone 11-26c
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was IgD.
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry