Please login to your online account to display your discounted pricing

Rat anti-IgA, Clone: mA-6E1, PE, Novus Biologicals™

$192.00 - $386.00

Specifications

Antigen IgA
Clone mA-6E1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was IgA.
Gene ID (Entrez) 3493
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14378201

 
novus biologicals canada
NBP14378201MG
0.1mg Each for $289.00

NBP14378202

 
novus biologicals canada
NBP14378202MG
0.2mg Each for $386.00

NBP14378205

 
novus biologicals canada
NBP143782005MG
0.05mg Each for $192.00
Description & Specifications

Specifications

Antigen IgA
Clone mA-6E1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was IgA.
Gene ID (Entrez) 3493
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry