Please login to your online account to display your discounted pricing

Rat anti-IgA, Clone: mA-6E1, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143782005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14378205

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Antigen IgA
Format Affinity Purified
Host Species Rat
Applications Flow Cytometry
Immunogen The immunogen for this antibody was IgA.
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Clone mA-6E1
Conjugate PE
Formulation Phosphate buffer pH 7.2
Isotype IgG1, κ
Primary or Secondary Primary
Gene ID (Entrez) 3493
Concentration 0.2mg/mL