Please login to your online account to display your discounted pricing

Rat anti-IgA, Clone: mA-6E1, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14378202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14378202

  • / Each
 

Description & Specifications

Specifications

Antigen IgA
Format Affinity Purified
Applications Flow Cytometry
Quantity 0.2mg
Primary or Secondary Primary
Cross Reactivity Mouse
Formulation Phosphate buffer pH 7.2
Host Species Rat
Immunogen The immunogen for this antibody was IgA.
Isotype IgG1, κ
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3493
Clone mA-6E1
Concentration 0.2mg/mL
Conjugate PE