Please login to your online account to display your discounted pricing

Rat anti-IgA, Clone: mA-6E1, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14378201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14378201

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Monoclonal or Polyclonal Monoclonal
Immunogen The immunogen for this antibody was IgA.
Quantity 0.1mg
Conjugate PE
Format Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Purification Method Affinity Purified
Clone mA-6E1
Concentration 0.2mg/mL
Applications Flow Cytometry
Formulation Phosphate buffer pH 7.2
Host Species Rat
Primary or Secondary Primary
Gene ID (Entrez) 3493
Isotype IgG1, κ
Regulatory Status RUO
Antigen IgA