Please login to your online account to display your discounted pricing

Rat anti-IgA, Clone: mA-6E1, FITC, Novus Biologicals™

$175.00 - $362.00

Specifications

Antigen IgA
Clone mA-6E1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was IgA.
Gene ID (Entrez) 3493
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14396101

 
novus biologicals canada
NBP14396101MG
0.1mg Each for $252.00

NBP14396102

 
novus biologicals canada
NBP14396102MG
0.2mg Each for $362.00

NBP14396105

 
novus biologicals canada
NBP143961005MG
0.05mg Each for $175.00
Description & Specifications

Specifications

Antigen IgA
Clone mA-6E1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was IgA.
Gene ID (Entrez) 3493
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG1, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry