Please login to your online account to display your discounted pricing

Rat anti-ICOS, Clone: 7E.17G9, PE, Novus Biologicals™

$197.00 - $296.00

Specifications

Antigen ICOS
Clone 7E.17G9
Immunogen The immunogen for this antibody was ICOS.
Gene ID (Entrez) 29851
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14370801

 
novus biologicals canada
NBP14370801MG
0.1mg Each for $296.00

NBP14370805

 
novus biologicals canada
NBP143708005MG
0.05mg Each for $197.00
Description & Specifications

Specifications

Antigen ICOS
Clone 7E.17G9
Immunogen The immunogen for this antibody was ICOS.
Gene ID (Entrez) 29851
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry