Please login to your online account to display your discounted pricing

Rat anti-ICOS, Clone: 7E.17G9, Biotin, Novus Biologicals™

$252.00 - $725.00

Specifications

Antigen ICOS
Clone 7E.17G9
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was ICOS.
Gene ID (Entrez) 29851
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP1434735

 
novus biologicals canada
NBP14347305MG
0.5mg Each for $725.00

NBP1434735M

 
novus biologicals canada
NBP143473005MG
0.05mg Each for $252.00
Description & Specifications

Specifications

Antigen ICOS
Clone 7E.17G9
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was ICOS.
Gene ID (Entrez) 29851
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry