Please login to your online account to display your discounted pricing

Rat anti-ICAM-2/CD102, Clone: 3C4, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen ICAM-2/CD102
Clone 3C4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was ICAM2.
Gene ID (Entrez) 3384
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14337201

 
novus biologicals canada
NBP14337201MG
0.1mg Each for $285.00

NBP14337203

 
novus biologicals canada
NBP1433720025MG
0.025mg Each for $219.00
Description & Specifications

Specifications

Antigen ICAM-2/CD102
Clone 3C4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was ICAM2.
Gene ID (Entrez) 3384
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation