Please login to your online account to display your discounted pricing

Rat anti-ICAM-2/CD102, Clone: 3C4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14337201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14337201

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Format Affinity Purified
Formulation Aqueous buffer (contains stabilizer if necessary)
Host Species Rat
Immunogen The immunogen for this antibody was ICAM2.
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3384
Conjugate Unlabeled
Cross Reactivity Mouse
Antigen ICAM-2/CD102
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Clone 3C4
Concentration 0.5mg/mL