Please login to your online account to display your discounted pricing

Rat anti-ICAM-2/CD102, Clone: 3C4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1433720025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14337203

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Formulation Aqueous buffer (contains stabilizer if necessary)
Host Species Rat
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Purification Method Affinity Purified
Quantity 0.025mg
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Conjugate Unlabeled
Antigen ICAM-2/CD102
Format Affinity Purified
Immunogen The immunogen for this antibody was ICAM2.
Isotype IgG2a, κ
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3384
Clone 3C4
Concentration 0.5mg/mL
Regulatory Status RUO