missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-HSP90 alpha, Clone: 9D2, Novus Biologicals™

$309.05 - $517.67

Specifications

Antigen HSP90 alpha
Clone 9D2
Immunogen Purified Hsp90 isolated from human therapeutic orchiectomy specimens.
Gene ID (Entrez) 3320
Formulation Liquid. In PBS, pH 7.2 containing 50% glycerol. Protein G purified.
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP19750802 Novus Biologicals Canada
NBP19750802MG
0.2mg Each for $517.67
NBP19750805 Novus Biologicals Canada
NBP197508005MG
0.05mg Each for $309.05
Specifications Description & Specifications

Specifications

HSP90 alpha
Purified Hsp90 isolated from human therapeutic orchiectomy specimens.
Liquid. In PBS, pH 7.2 containing 50% glycerol. Protein G purified.
Primary
Rat
Unlabeled
Purified
9D2
3320
Monoclonal
Store at -20°C. Avoid freeze-thaw cycles.
IgG2a
Purified
Chromatin Immunoprecipitation,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot