Please login to your online account to display your discounted pricing

Rat anti-HSP90 alpha, Clone: 9D2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP197508005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19750805

  • / Each
 

Description & Specifications

Specifications

Applications Chromatin Immunoprecipitation
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3320
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Human
Format Purified
Formulation Liquid. In PBS, pH 7.2 containing 50% glycerol. Protein G purified.
Immunogen Purified Hsp90 isolated from human therapeutic orchiectomy specimens.
Quantity 0.05mg
Antigen HSP90 alpha
Host Species Rat
Isotype IgG2a
Purification Method Purified
Primary or Secondary Primary
Clone 9D2