Please login to your online account to display your discounted pricing

Rat anti-HSP90 alpha, Clone: 9D2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP19750802MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19750802

  • / Each
 

Description & Specifications

Specifications

Applications Chromatin Immunoprecipitation
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Formulation Liquid. In PBS, pH 7.2 containing 50% glycerol. Protein G purified.
Primary or Secondary Primary
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene ID (Entrez) 3320
Conjugate Unlabeled
Host Species Rat
Immunogen Purified Hsp90 isolated from human therapeutic orchiectomy specimens.
Antigen HSP90 alpha
Quantity 0.2mg
Format Purified
Isotype IgG2a
Cross Reactivity Chicken
Cross Reactivity Human
Purification Method Purified
Clone 9D2
Regulatory Status RUO