Please login to your online account to display your discounted pricing

Rat anti-GM-CSF R alpha, Clone: 12-3A3-1B10, PE, Novus Biologicals™

$197.00 - $395.00

Specifications

Antigen GM-CSF R alpha
Clone 12-3A3-1B10
Immunogen The immunogen for this antibody is GM-CSF Receptor alpha.
Gene ID (Entrez) 1438
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200324

 
novus biologicals canada
NBP200324100TESTS
100 Tests Each for $395.00

NBP20032425

 
novus biologicals canada
NBP20032425TESTS
25 Tests Each for $197.00
Description & Specifications

Specifications

Antigen GM-CSF R alpha
Clone 12-3A3-1B10
Immunogen The immunogen for this antibody is GM-CSF Receptor alpha.
Gene ID (Entrez) 1438
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Human
Isotype IgG1
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry