Please login to your online account to display your discounted pricing

Rat anti-GM-CSF, Clone: SPM469, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen GM-CSF
Clone SPM469
Concentration 0.2mg/mL
Immunogen Recombinant human GM-CSF protein
Gene Accession No. P04141
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23296801

 
novus biologicals canada
NBP23296801ML
0.1mL Each for $139.00

NBP23296810

 
novus biologicals canada
NBP23296810ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen GM-CSF
Clone SPM469
Concentration 0.2mg/mL
Immunogen Recombinant human GM-CSF protein
Gene Accession No. P04141
Gene ID (Entrez) 1437
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Rat
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Primate
Cross Reactivity Rhesus Ma
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting