Please login to your online account to display your discounted pricing

Rat anti-GM-CSF, Clone: SPM469, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23296810ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23296810

  • / Each
 

Description & Specifications

Specifications

Antigen GM-CSF
Host Species Rat
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Primate
Cross Reactivity Rhesus Ma
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P04141
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant human GM-CSF protein
Isotype IgG2a, κ
Purification Method Purified
Primary or Secondary Primary
Gene ID (Entrez) 1437
Reconstitution Protein A purified
Clone SPM469
Concentration 0.2mg/mL
Conjugate Unlabeled