Please login to your online account to display your discounted pricing

Rat anti-GM-CSF, Clone: SPM469, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23296801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23296801

  • / Each
 

Description & Specifications

Specifications

Antigen GM-CSF
Formulation PBS with 0.05% BSA.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2a, κ
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Primate
Cross Reactivity Rhesus Ma
Format Purified
Gene Accession No. P04141
Host Species Rat
Immunogen Recombinant human GM-CSF protein
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 1437
Reconstitution Protein A purified
Clone SPM469
Concentration 0.2mg/mL