missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-GM-CSF, Clone: SPM469, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen GM-CSF
Clone SPM469
Concentration 1mg/mL
Immunogen Recombinant human GM-CSF protein
Gene Accession No. P04141
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23477401 Novus Biologicals Canada
NBP23477401MG
0.1mg Each for $384.00
NBP23477402 Novus Biologicals Canada
NBP23477402MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

GM-CSF
1mg/mL
P04141
10mM PBS. Does not contain BSA. Azide Free.
Primary
Rat
Unlabeled
Purified
SPM469
Recombinant human GM-CSF protein
1437
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2a, κ
Purified
Blocking,ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunoprecipitation,Neutralization,Western Blot