Please login to your online account to display your discounted pricing

Rat anti-GM-CSF, Clone: SPM469, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23477402MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23477402

  • / Each
 

Description & Specifications

Specifications

Antigen GM-CSF
Applications Blocking
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Neutralization
Applications Western Blot
Clone SPM469
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P04141
Host Species Rat
Immunogen Recombinant human GM-CSF protein
Isotype IgG2a, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1437
Reconstitution Protein A purified
Target Species Cynomolgus
Target Species Human
Target Species Monkey
Target Species Primate
Target Species Rhesus