Please login to your online account to display your discounted pricing

Rat anti-GM-CSF, Clone: BVD2-21C11, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23457001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23457001

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Primate
Cross Reactivity Rhesus Ma
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Gene ID (Entrez) 1437
Host Species Rat
Conjugate Unlabeled
Quantity 0.1mg
Applications Blocking
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Neutralization
Applications Western Blotting
Isotype IgG2a, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Format Purified
Gene Accession No. P04141
Concentration 1mg/mL
Reconstitution Protein A purified
Antigen GM-CSF
Immunogen Recombinant human GM-CSF protein
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Regulatory Status RUO
Purification Method Purified
Clone BVD2-21C11