Please login to your online account to display your discounted pricing

Rat anti-GL7 (T and B Cell Activation Marker), Clone: GL-7, Biotin, Novus Biologicals™

$329.00 - $725.00

Specifications

Antigen GL7 (T and B Cell Activation Marker)
Clone GL-7
Concentration 0.5mg/mL
Immunogen Ly-77).
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20036110

 
novus biologicals canada
NBP200361100UG
100μg Each for $329.00

NBP20036155

 
novus biologicals canada
NBP200361500UG
500μg Each for $725.00
Description & Specifications

Specifications

Antigen GL7 (T and B Cell Activation Marker)
Clone GL-7
Concentration 0.5mg/mL
Immunogen Ly-77).
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgM
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry