Please login to your online account to display your discounted pricing

Rat anti-GATA-4, Clone: Evan, Novus Biologicals™

$274.00 - $362.00

Specifications

Antigen GATA-4
Clone Evan
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Gata4.
Gene ID (Entrez) 2626
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14311201

 
novus biologicals canada
NBP14311201MG
0.1mg Each for $362.00

NBP14311203

 
novus biologicals canada
NBP1431120025MG
0.025mg Each for $274.00
Description & Specifications

Specifications

Antigen GATA-4
Clone Evan
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Gata4.
Gene ID (Entrez) 2626
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Western Blot