Please login to your online account to display your discounted pricing

Rat anti-GATA-4, Clone: Evan, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1431120025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14311203

  • / Each
 

Description & Specifications

Specifications

Antigen GATA-4
Applications Western Blot
Clone Evan
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Rat
Immunogen The immunogen for this antibody was Gata4.
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2626
Target Species Human
Target Species Murine