Please login to your online account to display your discounted pricing

Rat anti-GATA-4, Clone: Evan, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14311201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14311201

  • / Each
 

Description & Specifications

Specifications

Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Clone Evan
Formulation PBS [pH 7.2]
Immunogen The immunogen for this antibody was Gata4.
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 0.1mg
Gene ID (Entrez) 2626
Conjugate Unlabeled
Antigen GATA-4
Primary or Secondary Primary
Concentration 0.5mg/mL