Please login to your online account to display your discounted pricing

Rat anti-GATA-3, Clone: TWAJ, PE, Novus Biologicals™

$175.00 - $362.00

Specifications

Antigen GATA-3
Clone TWAJ
Immunogen The immunogen for this antibody is Gata-3 (Gata3).
Gene ID (Entrez) 2625
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200287

 
novus biologicals canada
NBP200287100TESTS
100 Tests Each for $362.00

NBP20028725

 
novus biologicals canada
NBP20028725TESTS
25 Tests Each for $175.00
Description & Specifications

Specifications

Antigen GATA-3
Clone TWAJ
Immunogen The immunogen for this antibody is Gata-3 (Gata3).
Gene ID (Entrez) 2625
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry