Please login to your online account to display your discounted pricing

Rat anti-Galectin-3, Clone: M3/38, Novus Biologicals™

$252.00 - $384.00

Specifications

Antigen Galectin-3
Clone M3/38
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Galectin3 Mac2.
Gene ID (Entrez) 3958
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14331301

 
novus biologicals canada
NBP14331301MG
0.1mg Each for $329.00

NBP14331305

 
novus biologicals canada
NBP143313005MG
0.05mg Each for $252.00

NBP1433135

 
novus biologicals canada
NBP14331305MG
0.5mg Each for $384.00
Description & Specifications

Specifications

Antigen Galectin-3
Clone M3/38
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Galectin3 Mac2.
Gene ID (Entrez) 3958
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting