Please login to your online account to display your discounted pricing

Rat anti-Galectin-3, Clone: M3/38, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143313005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14331305

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Host Species Rat
Antigen Galectin-3
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Quantity 0.05mg
Isotype IgG2a, κ
Conjugate Unlabeled
Immunogen The immunogen for this antibody was Galectin3 Mac2.
Storage Requirements Store at 4°C. Do not freeze.
Format Affinity Purified
Regulatory Status RUO
Gene ID (Entrez) 3958
Clone M3/38
Purification Method Affinity Purified
Concentration 0.5mg/mL