Please login to your online account to display your discounted pricing

Rat anti-Galectin-3, Clone: M3/38, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14331301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14331301

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mg
Primary or Secondary Primary
Gene ID (Entrez) 3958
Cross Reactivity Mouse
Antigen Galectin-3
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Rat
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen The immunogen for this antibody was Galectin3 Mac2.
Isotype IgG2a, κ
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Clone M3/38
Concentration 0.5mg/mL
Conjugate Unlabeled