missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-Galectin-3, Clone: M3/38, Biotin, Novus Biologicals™

$131.00 - $472.00

Specifications

Antigen Galectin-3
Clone M3/38
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Galectin3 Mac2.
Gene ID (Entrez) 3958
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14355301 Novus Biologicals Canada
NBP14355301MG
0.1mg Each for $252.00
NBP14355305 Novus Biologicals Canada
NBP143553005MG
0.05mg Each for $131.00
NBP1435535 Novus Biologicals Canada
NBP14355305MG
0.5mg Each for $472.00
Specifications Description & Specifications

Specifications

Galectin-3
0.5mg/mL
3958
Monoclonal
Store at 4°C. Do not freeze.
IgG2a, κ
Affinity Purified
Flow Cytometry
M3/38
The immunogen for this antibody was Galectin3 Mac2.
PBS [pH 7.2]
Primary
Rat
Biotin
Affinity Purified