missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-Flt-3/Flk-2/CD135, Clone: A2F10, Novus Biologicals™

$219.00 - $362.00

Specifications

Antigen Flt-3/Flk-2/CD135
Clone A2F10
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD135.
Gene ID (Entrez) 2322
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14335201 Novus Biologicals Canada
NBP14335201MG
0.1mg Each for $285.00
NBP14335205 Novus Biologicals Canada
NBP143352005MG
0.05mg Each for $219.00
NBP1433525 Novus Biologicals Canada
NBP14335205MG
0.5mg Each for $362.00
Specifications Description & Specifications

Specifications

Flt-3/Flk-2/CD135
0.5mg/mL
2322
Monoclonal
Store at 4°C. Do not freeze.
IgG2a, κ
Affinity Purified
Flow Cytometry,Functional Assay,Immunoprecipitation
A2F10
The immunogen for this antibody was CD135.
Phosphate buffer pH 7.2
Primary
Rat
Unlabeled
Affinity Purified