Please login to your online account to display your discounted pricing

Rat anti-Endoglin/CD105, Clone: MJ7/18, Biotin, Novus Biologicals™

$175.00 - $505.00

Specifications

Antigen Endoglin/CD105
Clone MJ7/18
Immunogen inflamed mouse skin
Gene ID (Entrez) 2022
Formulation Phosphate buffer pH 7.2. Immunogen affinity purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14359801

 
novus biologicals canada
NBP14359801MG
0.1mg Each for $252.00

NBP14359805

 
novus biologicals canada
NBP143598005MG
0.05mg Each for $175.00

NBP1435985

 
novus biologicals canada
NBP14359805MG
0.5mg Each for $505.00
Description & Specifications

Specifications

Antigen Endoglin/CD105
Clone MJ7/18
Immunogen inflamed mouse skin
Gene ID (Entrez) 2022
Formulation Phosphate buffer pH 7.2. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry