Please login to your online account to display your discounted pricing

Rat anti-EMMPRIN/CD147, Clone: RL73, Novus Biologicals™

$214.00 - $278.00

Specifications

Antigen EMMPRIN/CD147
Clone RL73
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD147.
Gene ID (Entrez) 682
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14334501

 
novus biologicals canada
NBP14334501MG
0.1mg Each for $278.00

NBP14334505

 
novus biologicals canada
NBP143345005MG
0.05mg Each for $214.00
Description & Specifications

Specifications

Antigen EMMPRIN/CD147
Clone RL73
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD147.
Gene ID (Entrez) 682
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunoprecipitation