Please login to your online account to display your discounted pricing

Rat anti-EMMPRIN/CD147, Clone: RL73, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14334501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14334501

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunoprecipitation
Host Species Rat
Cross Reactivity Mouse
Antigen EMMPRIN/CD147
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Immunogen The immunogen for this antibody was CD147.
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 682
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 0.1mg
Clone RL73
Concentration 0.2mg/mL
Conjugate Unlabeled