Please login to your online account to display your discounted pricing

Rat anti-EMMPRIN/CD147, Clone: RL73, FITC, Novus Biologicals™

$171.00 - $536.00

Specifications

Antigen EMMPRIN/CD147
Clone RL73
Immunogen The immunogen for this antibody was CD147.
Gene ID (Entrez) 682
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14397001

 
novus biologicals canada
NBP14397001MG
0.1mg Each for $246.00

NBP1439705

 
novus biologicals canada
NBP14397005MG
0.5mg Each for $536.00

NBP14397005

 
novus biologicals canada
NBP143970005MG
0.05mg Each for $171.00
Description & Specifications

Specifications

Antigen EMMPRIN/CD147
Clone RL73
Immunogen The immunogen for this antibody was CD147.
Gene ID (Entrez) 682
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry