Please login to your online account to display your discounted pricing

Rat anti-EMMPRIN/CD147, Clone: RL73, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14397005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1439705

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Cross Reactivity Mouse
Format Affinity Purified
Immunogen The immunogen for this antibody was CD147.
Quantity 0.5mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 682
Conjugate FITC
Antigen EMMPRIN/CD147
Formulation Phosphate buffer pH 7.2
Host Species Rat
Isotype IgG2a, κ
Purification Method Affinity Purified
Monoclonal or Polyclonal Monoclonal
Clone RL73
Primary or Secondary Primary