Please login to your online account to display your discounted pricing

Rat anti-EMMPRIN/CD147, Clone: RL73, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143970005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14397005

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Formulation Phosphate buffer pH 7.2
Format Affinity Purified
Isotype IgG2a, κ
Immunogen The immunogen for this antibody was CD147.
Cross Reactivity Mouse
Regulatory Status RUO
Conjugate FITC
Clone RL73
Antigen EMMPRIN/CD147
Quantity 0.05mg
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 682
Purification Method Affinity Purified
Host Species Rat
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.