Please login to your online account to display your discounted pricing

Rat anti-EMMPRIN/CD147, Clone: RL73, FITC, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP14397001MG

 View all options for this product


For Research Use Only

Catalog No. NBP14397001


Description & Specifications


Antigen EMMPRIN/CD147
Applications Flow Cytometry
Clone RL73
Conjugate FITC
Cross Reactivity Mouse
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Rat
Immunogen The immunogen for this antibody was CD147.
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 682