Please login to your online account to display your discounted pricing

Rat anti-Dendritic Cell Marker, Clone: 33D1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14328401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14328401

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Quantity 0.1mg
Primary or Secondary Primary
Antigen Dendritic Cell Marker
Host Species Rat
Conjugate Unlabeled
Immunogen The immunogen for this antibody was Dendritic Cell Marker.
Format Affinity Purified
Regulatory Status RUO
Cross Reactivity Mouse
Clone 33D1
Concentration 0.5mg/mL
Purification Method Affinity Purified
Formulation Phosphate buffer pH 7.2
Isotype IgG2b, κ
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal