Please login to your online account to display your discounted pricing

Rat anti-Dendritic Cell Marker, Clone: 33D1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143284005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14328405

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Format Affinity Purified
Immunogen The immunogen for this antibody was Dendritic Cell Marker.
Primary or Secondary Primary
Cross Reactivity Mouse
Antigen Dendritic Cell Marker
Formulation Phosphate buffer pH 7.2
Host Species Rat
Isotype IgG2b, κ
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Clone 33D1
Concentration 0.5mg/mL
Quantity 0.05mg
Conjugate Unlabeled