Please login to your online account to display your discounted pricing

Rat anti-Dendritic Cell Marker, Clone: 33D1, Biotin, Novus Biologicals™

$252.00 - $725.00

Specifications

Antigen Dendritic Cell Marker
Clone 33D1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Dendritic Cell Marker.
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14352901

 
novus biologicals canada
NBP14352901MG
0.1mg Each for $329.00

NBP14352905

 
novus biologicals canada
NBP143529005MG
0.05mg Each for $252.00

NBP1435295

 
novus biologicals canada
NBP14352905MG
0.5mg Each for $725.00
Description & Specifications

Specifications

Antigen Dendritic Cell Marker
Clone 33D1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Dendritic Cell Marker.
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry