missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-DC-SIGN/CD209, Clone: LWC06, Novus Biologicals™

$274.00 - $406.00

Specifications

Antigen DC-SIGN/CD209
Clone LWC06
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CIRE.
Gene ID (Entrez) 30835
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14332801 Novus Biologicals Canada
NBP14332801MG
0.1mg Each for $406.00
NBP14332803 Novus Biologicals Canada
NBP1433280025MG
0.025mg Each for $274.00
Specifications Description & Specifications

Specifications

DC-SIGN/CD209
0.5mg/mL
30835
Monoclonal
Store at 4°C. Do not freeze.
IgG2a
Affinity Purified
Immunoprecipitation,Western Blot
LWC06
The immunogen for this antibody was CIRE.
Phosphate buffer pH 7.2
Primary
Rat
Unlabeled
Affinity Purified